Alfuzosin hydrochloride ≥98.0% (by HPLC)
Proveedor: TCI
Sinónimos:
N-[3-[(4-Amino-6,7-dimethoxy-2-quinazolinyl)(methyl)amino]propyl]tetrahydro-2-furancarboxamide Hydrochloride, 4-Amino-6,7-dimethoxy-2-[methyl[3-[(tetrahydro-2-furoyl)amino]propyl]amino]quinazoline Hydrochloride, Alfuzosin HCl, N-(3-((4-Amino-6,7-dimethoxyquinazolin-2-yl)(methyl)amino)propyl)tetrahydrofuran-2-carboxamide hydrochloride
A2591-100MG
A2591-1G
TCIAA2591-100MGEA
16
EUR
TCIAA2591-100MG
TCIAA2591-1G
Alfuzosin hydrochloride ≥98.0% (by HPLC)
Alfuzosin hydrochloride
Formula:
C₁₉H₂₇N₅O₄ MW: 425,91 g/mol |
Número MDL:
MFCD00879135 Núm. CAS: 81403-68-1 Índice Merck: 14,00238 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...