(Acetylthio)acetic acid ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-(Acetylthio)acetic acid
A2621-5G
A2621-25G
TCIAA2621-5GEA
93
EUR
TCIAA2621-5G
TCIAA2621-25G
(Acetylthio)acetic acid ≥98.0% (by GC, titration analysis)
(Acetylthio)acetic acid
Formula:
C₄H₆O₃S MW: 134,15 g/mol Pto. Ebullición: 160°C |
Número MDL:
MFCD00021763 Núm. CAS: 1190-93-8 EINECS: 214-725-7 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...