2-Amino-9,9-dimethylfluorene ≥99.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
9,9-Dimethyl-9H-fluoren-2-amine
A2634-200MG
A2634-1G
TCIAA2634-200MGEA
16
EUR
TCIAA2634-200MG
TCIAA2634-1G
2-Amino-9,9-dimethylfluorene ≥99.0% (by HPLC, titration analysis)
2-Amino-9,9-dimethylfluorene
Formula:
C₁₅H₁₅N MW: 209,29 g/mol |
Número MDL:
MFCD09953790 Núm. CAS: 108714-73-4 |
Resultados test especificaciones
Appearance | White to Light yellow to Green powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 166.0 to 170.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...