(R)-3-Amino-3-phenylpropionic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: H-D-β-Phenyl-Ala-OH, 3-Amino-3-phenylpropionic-acid, (R)-3-Amino-3-phenylpropanoic acid, (R)-b-Amino-benzenepropanoic acid

A2655-200MG A2655-1G
TCIAA2655-200MGEA 30 EUR
TCIAA2655-200MG TCIAA2655-1G
(R)-3-Amino-3-phenylpropionic acid ≥98.0% (by HPLC, titration analysis)
(R)-3-Amino-3-phenylpropionic acid
Formula: C₉H₁₁NO₂
MW: 165,189 g/mol
Número MDL: MFCD01076237
Núm. CAS: 13921-90-9

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +6.0 to +8.0 deg (C=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR