N-Allyl-p-toluenesulphonamide ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
Sinónimos:
N-Allyl-4-methylbenzenesulphonamide
A2665-1G
A2665-5G
TCIAA2665-1GEA
68
EUR
TCIAA2665-1G
TCIAA2665-5G
N-Allyl-p-toluenesulphonamide ≥98.0% (por GC)
N-Allyl-p-toluenesulphonamide
Formula:
C₁₀H₁₃NO₂S MW: 211,28 g/mol |
Número MDL:
MFCD00182633 Núm. CAS: 50487-71-3 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 61.0 to 65.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...