3-Acetyl-2-(aminosulphonyl)-5-chlorothiophene ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
3-Acetyl-5-chloro-2-thiophenesulphonamide, 3-Acetyl-5-chlorothiophene-2-sulphonamide
A2667-1G
A2667-5G
TCIAA2667-1GEA
134
EUR
TCIAA2667-1G
TCIAA2667-5G
3-Acetyl-2-(aminosulphonyl)-5-chlorothiophene ≥98.0% (by HPLC, titration analysis)
3-Acetyl-2-(aminosulphonyl)-5-chlorothiophene
Formula:
C₆H₆ClNO₃S₂ MW: 239,69 g/mol |
Número MDL:
MFCD09033309 Núm. CAS: 160982-10-5 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...