6-Acetoxy-7-methoxy-3H-quinazolin-4-one ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
6-Acetoxy-7-methoxy-4-quinazolinol, 6-Acetoxy-7-methoxy-4(3H)-quinazolinone, 6-Acetoxy-7-methoxy-3,4-dihydroquinazolin-4-one, 6-Acetoxy-7-methoxy-4-hydroxyquinazoline
A2717-1G
A2717-5G
TCIAA2717-1GEA
25
EUR
TCIAA2717-1G
TCIAA2717-5G
6-Acetoxy-7-methoxy-3H-quinazolin-4-one ≥98.0% (by HPLC, titration analysis)
6-Acetoxy-7-methoxy-3H-quinazolin-4-one
Formula:
C₁₁H₁₀N₂O₄ MW: 234,21 g/mol |
Número MDL:
MFCD09951978 Núm. CAS: 179688-53-0 |
Resultados test especificaciones
Appearance | White to Yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...