4-(Methylsulphonyl)aniline ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
A2725-25G
A2725-100G
TCIAA2725-25GEA
39
EUR
TCIAA2725-25G
TCIAA2725-100G
4-(Methylsulphonyl)aniline ≥98.0% (by GC, titration analysis)
4-(Methylsulphonyl)aniline
Formula:
C₇H₉NO₂S MW: 171,21 g/mol |
Número MDL:
MFCD00025363 Núm. CAS: 5470-49-5 EINECS: 226-803-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 135.0 to 139.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...