2-Amino-4'-fluorobenzophenone ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: (2-Aminophenyl)(4-fluorophenyl)methanone

A2760-1G A2760-5G
TCIAA2760-1GEA 23 EUR
TCIAA2760-1G TCIAA2760-5G
2-Amino-4'-fluorobenzophenone ≥98.0% (by GC, titration analysis)
2-Amino-4'-fluorobenzophenone
Formula: C₁₃H₁₀FNO
MW: 215,2230 g/mol
Número MDL: MFCD06658166
Núm. CAS: 3800-06-4

Order Now

Resultados test especificaciones

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 127.0 to 131.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR