Azadibenzocyclooctyne-amine ≥95.0% (by HPLC)

Proveedor: TCI

Sinónimos: DBCO-amine, 3-Amino-1-(11,12-didehydrodibenz[b,f]azocin-5(6H)-yl)-1-propanone, Dibenzocyclooctyne-amine

A2763-25MG A2763-100MG
TCIAA2763-25MGEA 101 EUR
TCIAA2763-25MG TCIAA2763-100MG
Azadibenzocyclooctyne-amine ≥95.0% (by HPLC)
Azadibenzocyclooctyne-amine

This product is unavailable in the U.S.

Formula: C₁₈H₁₆N₂O
MW: 276,33 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD22380759
Núm. CAS: 1255942-06-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b220908f63888e6aa22a7ed485bc5eebb0ee72e&vid=6b220908f63888e6aa22a7ed485bc5eebb0ee72e&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Brown powder to crystal
Purity (HPLC) min. 95.0 area%
Elemental analysis (Nitrogen) 9.50 to 10.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR