2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

A2773-1G A2773-5G
TCIAA2773-1GEA 16 EUR
TCIAA2773-1G TCIAA2773-5G
2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide ≥98.0% (by GC, titration analysis)
2-Amino-N-(2-chloro-6-methylphenyl)thiazole-5-carboxamide
Formula: C₁₁H₁₀ClN₃OS
MW: 267,73 g/mol
Número MDL: MFCD10000630
Núm. CAS: 302964-24-5

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 208.0 to 212.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR