5-Acetyl-2-chloropyridine ≥98.0% (por GC)
Proveedor: TCI
|
Atención
|
A2796-1G
A2796-5G
TCIAA2796-1GEA
26
EUR
TCIAA2796-1G
TCIAA2796-5G
5-Acetyl-2-chloropyridine ≥98.0% (por GC)
5-Acetyl-2-chloropyridine
Formula:
C₇H₆ClNO MW: 155,5816 g/mol |
Número MDL:
MFCD03407343 Núm. CAS: 55676-22-7 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...