7-Amino-2-benzoxazolinone ≥95.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
7-Amino-2-hydroxybenzoxazole, 7-Amino-2-benzoxazolone, 7-Aminobenzo[d]oxazol-2(3H)-one
A2806-250MG
A2806-1G
TCIAA2806-250MGEA
77
EUR
TCIAA2806-250MG
TCIAA2806-1G
7-Amino-2-benzoxazolinone ≥95.0% (by GC, titration analysis)
7-Amino-2-benzoxazolinone
Formula:
C₇H₆N₂O₂ MW: 150,14 g/mol |
Número MDL:
MFCD09753616 Núm. CAS: 81282-60-2 EINECS: 462-910-8 |
Resultados test especificaciones
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...