4-Acetoxybenzeneboronic acid (contains varying amounts of Anhydride)

Proveedor: TCI
Atención

Sinónimos: 4-Acetoxybenzeneboronic acid, 4-Acetoxyphenylboronic acid, (4-Acetoxyphenyl)boronic acid

A2818-200MG A2818-1G
TCIAA2818-200MGEA 23 EUR
TCIAA2818-200MG TCIAA2818-1G
4-Acetoxybenzeneboronic acid (contains varying amounts of Anhydride)
4-Acetoxybenzeneboronic acid
Formula: C₈H₉BO₄
MW: 179.97 g/mol
Número MDL: MFCD09027198
Núm. CAS: 177490-82-3

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR