2-Amino-4-(4-chlorophenyl)thiazole ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
4-(4-Chlorophenyl)thiazol-2-amine
A2841-1G
A2841-5G
TCIAA2841-1GEA
39
EUR
TCIAA2841-1G
TCIAA2841-5G
2-Amino-4-(4-chlorophenyl)thiazole ≥98.0% (by GC, titration analysis)
2-Amino-4-(4-chlorophenyl)thiazole
Formula:
C₉H₇ClN₂S MW: 210,69 g/mol |
Número MDL:
MFCD00047058 Núm. CAS: 2103-99-3 UN: 2811 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 167.0 to 171.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...