2'-Amino-4',5'-dimethoxyacetophenone ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 1-(2-Amino-4,5-dimethoxyphenyl)ethanone

A2842-1G A2842-5G
TCIAA2842-1GEA 22 EUR
TCIAA2842-1G TCIAA2842-5G
2'-Amino-4',5'-dimethoxyacetophenone ≥98.0% (by GC, titration analysis)
2'-Amino-4',5'-dimethoxyacetophenone
Formula: C₁₀H₁₃NO₃
MW: 195,22 g/mol
Número MDL: MFCD00016646
Núm. CAS: 4101-30-8

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 105.0 to 109.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR