Ácido 6-metoxiantranilico ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

A2847-1G A2847-5G
TCIAA2847-1GEA 68 EUR
TCIAA2847-1G TCIAA2847-5G
Ácido 6-metoxiantranilico ≥98.0% (by HPLC, titration analysis)
Ácido 6-metoxiantranilico
Formula: C₈H₉NO₃
MW: 167,16 g/mol
Número MDL: MFCD01941328
Núm. CAS: 53600-33-2

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 86.0 to 90.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR