2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene ≥98.0% (by HPLC, total nitrogen)
Proveedor: TCI
A2858-1G
A2858-5G
TCIAA2858-1GEA
47
EUR
TCIAA2858-1G
TCIAA2858-5G
2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene ≥98.0% (by HPLC, total nitrogen)
2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene
Formula:
C₃₃H₂₆BrN |
Número MDL:
MFCD20040455 Núm. CAS: 1246562-40-2 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 159.0 to 163.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...