Abiraterone Acetate ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 3beta-17-(3-Pyridyl)androsta-5,16-dien-3-ol Acetate

A2891-200MG A2891-1G
TCIAA2891-200MGEA 52 EUR
TCIAA2891-200MG TCIAA2891-1G
Abiraterone Acetate ≥98.0% (by HPLC, titration analysis)
Abiraterone Acetate
Formula: C₂₆H₃₃NO₂
MW: 391.56 g/mol
Temperatura de almacenaje: Congelador
Número MDL: MFCD00934213
Núm. CAS: 154229-18-2

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 145.0 to 149.0 deg-C
Specific rotation [a]20/D -40.0 to -45.0 deg (C=2,EtOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR