Fmoc-Glu(OAll)-OH ≥96.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
N-Fmoc-L-glutamic acid 5-allyl ester, N-Fmoc-L-glutamic acid 5-Allyl ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamic acid 5-Allyl ester, Fmoc-Glu(allyl ester)-OH, 5-Allyl N-Fmoc-L-glutamate, Fmoc-Glu(OAll)-OH, 5-Allyl N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamate
A2895-1G
A2895-5G
TCIAA2895-1GEA
42
EUR
TCIAA2895-1G
TCIAA2895-5G
Fmoc-Glu(OAll)-OH ≥96.0% (by HPLC, titration analysis)
Fmoc-Glu(OAll)-OH
Formula:
C₂₃H₂₃NO₆ Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00190879 Núm. CAS: 133464-46-7 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystaline |
Purity (HPLC) | min. 96.0 area% |
Purity (Neutralization titration) | min. 96.0 % |
Melting point | 125.0 to 129.0 deg-C |
Specific rotation [a]20/D | -15.0 to -19.0 deg (C=1,DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...