2-Aminoisonicotinamide ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Peligro
|
Sinónimos:
2-Aminopyridine-4-carboxamide, 2-Amino-4-pyridinecarboxamide
A2899-1G
A2899-5G
TCIAA2899-1GEA
43
EUR
TCIAA2899-1G
TCIAA2899-5G
2-Aminoisonicotinamide ≥98.0% (by HPLC, titration analysis)
2-Aminoisonicotinamide
Formula:
C₆H₇N₃O MW: 137,14 g/mol |
Número MDL:
MFCD03791262 Núm. CAS: 13538-42-6 UN: 2811 ADR: 6.1,III |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...