2-Amino-3-bromo-5,6-dimethylpyridine ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
6-Amino-5-bromo-2,3-lutidine, 3-Bromo-5,6-dimethylpyridin-2-amine
A2925-1G
A2925-5G
TCIAA2925-1GEA
82
EUR
TCIAA2925-1G
TCIAA2925-5G
2-Amino-3-bromo-5,6-dimethylpyridine ≥98.0% (by GC, titration analysis)
2-Amino-3-bromo-5,6-dimethylpyridine
Formula:
C₇H₉BrN₂ MW: 201,07 g/mol |
Número MDL:
MFCD04039386 Núm. CAS: 161091-49-2 |
Resultados test especificaciones
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 114.0 to 118.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...