3'-Amino-2'-hydroxybiphenyl-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 3'-Amino-2'-hydroxybiphenyl-3-carboxylic Acid, 3'-Amino-2'-hydroxy-[1,1'-biphenyl]-3-carboxylic acid, 3-(3-Amino-2-hydroxyphenyl)benzoic acid

A2944-200MG A2944-1G
TCIAA2944-200MGEA 38 EUR
TCIAA2944-200MG TCIAA2944-1G
3'-Amino-2'-hydroxybiphenyl-3-carboxylic acid ≥98.0% (by HPLC, titration analysis)
3'-Amino-2'-hydroxybiphenyl-3-carboxylic acid
Formula: C₁₃H₁₁NO₃
MW: 229,24 g/mol
Número MDL: MFCD10001467
Núm. CAS: 376592-93-7

Order Now

Resultados test especificaciones

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR