Atipamezole hydrochloride ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI

Sinónimos: 4-(2-Ethyl-2,3-dihydro-1H-inden-2-yl)-1H-imidazole Hydrochloride, Atipamezole Hydrochloride, 4-(2-Ethyl-2,3-dihydro-1H-inden-2-yl)-1H-imidazole hydrochloride

A2956-25MG A2956-100MG
TCIAA2956-25MGEA 94 EUR
TCIAA2956-25MG TCIAA2956-100MG
Atipamezole hydrochloride ≥98.0% (by HPLC, titration analysis)
Atipamezole hydrochloride
Formula: C₁₄H₁₆N₂·HCl
MW: 248,75 g/mol
Número MDL: MFCD06407819
Núm. CAS: 104075-48-1
Índice Merck: 14,00861
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e044318f0cbf57aa767562a7413c7a386662e0ca&vid=e044318f0cbf57aa767562a7413c7a386662e0ca&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 218.0 to 222.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR