3-Amino-4-bromo-5-methylpyrazole ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
|
Atención
|
Sinónimos:
4-Bromo-5-methyl-1H-pyrazol-3-amine
A2970-1G
A2970-5G
TCIAA2970-1GEA
41
EUR
TCIAA2970-1G
TCIAA2970-5G
3-Amino-4-bromo-5-methylpyrazole ≥98.0% (by HPLC, titration analysis)
3-Amino-4-bromo-5-methylpyrazole
Formula:
C₄H₆BrN₃ MW: 176,02 g/mol |
Número MDL:
MFCD03412169 Núm. CAS: 1780-72-9 |
Resultados test especificaciones
Appearance | White to Brown powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 118.0 to 122.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...