2-Amino-5-bromo-4-methyl-3-nitropyridine ≥98.0% (by titrimetric analysis)
Proveedor: TCI
|
Atención
|
A2988-1G
A2988-5G
TCIAA2988-1GEA
42
EUR
TCIAA2988-1G
TCIAA2988-5G
2-Amino-5-bromo-4-methyl-3-nitropyridine ≥98.0% (by titrimetric analysis)
2-Amino-5-bromo-4-methyl-3-nitropyridine
Formula:
C₆H₆BrN₃O₂ MW: 232,0400 g/mol |
Número MDL:
MFCD00092197 Núm. CAS: 100367-40-6 |
Resultados test especificaciones
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 168.0 to 172.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...