Alcohol 3-amino-4-metilbencilico ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 3-Amino-4-methylbenzyl Alcohol

A2998-1G A2998-5G
TCIAA2998-1GEA 21 EUR
TCIAA2998-1G TCIAA2998-5G
Alcohol 3-amino-4-metilbencilico ≥98.0% (by GC, titration analysis)
3-Amino-4-methylbenzyl alcohol
Formula: C₈H₁₁NO
MW: 137,18 g/mol
Número MDL: MFCD00075051
Núm. CAS: 81863-45-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c0a10888275a1fa6a482ae3a8f2effffcb94a66&vid=1c0a10888275a1fa6a482ae3a8f2effffcb94a66&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR