2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride ≥98.0% (by HPLC) (contains varying amounts of Anhydride)
Proveedor: TCI
|
Atención
|
Sinónimos:
2-Amino-4-methoxycarbonyl phenylboronic acid Hydrochloride, 2-Amino-4-(methoxycarbonyl)phenylboronic acid hydrochloride, 2-Amino-4-(methoxycarbonyl)benzeneboronic acid hydrochloride, 2-Amino-4-methoxycarbonylphenylboronic acid HCl, (2-Amino-4-(methoxycarbonyl)phenyl)boronic acid hydrochloride, 2-Amino-4-(methoxycarbonyl)benzeneboronic acid Hydrochloride
Formula:
C₈H₁₀BNO₄·HCl MW: 231,44 g/mol Pto. de fusión: 180°C |
Número MDL:
MFCD02258941 Núm. CAS: 380430-55-7 |
Resultados test especificaciones
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | 97.0 to 108.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...