3-(4-Methoxyphenyl)-1-phenyl-1H-pyrazol-5-amine ≥98.0% (by HPLC, titration analysis)

Proveedor: TCI
Atención

Sinónimos: 5-Amino-3-(4-methoxyphenyl)-1-phenylpyrazole

A3055-1G A3055-5G
TCIAA3055-1GEA 58 EUR
TCIAA3055-1G TCIAA3055-5G
3-(4-Methoxyphenyl)-1-phenyl-1H-pyrazol-5-amine ≥98.0% (by HPLC, titration analysis)
3-(4-Methoxyphenyl)-1-phenyl-1H-pyrazol-5-amine
Formula: C₁₆H₁₅N₃O
Número MDL: MFCD00197061
Núm. CAS: 19652-13-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4ce76df43d8a15f90e17a65161d4214aa9b9ba6&vid=a4ce76df43d8a15f90e17a65161d4214aa9b9ba6&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Gray to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 188.0 to 192.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR