H-Phg(4-Cl)-OH ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
Sinónimos:
H-Phg(4-Cl)-OH, 4-Chloro-L-2-phenylglycine, L-2-(4-Chlorophenyl)glycine, (S)-2-Amino-2-(4-chlorophenyl)acetic acid
A3077-1G
A3077-5G
TCIAA3077-1GEA
35
EUR
TCIAA3077-1G
TCIAA3077-5G
H-Phg(4-Cl)-OH ≥98.0% (by HPLC, titration analysis)
H-Phg(4-Cl)-OH
Formula:
C₈H₈ClNO₂ MW: 185,6 g/mol |
Número MDL:
MFCD03701467 Núm. CAS: 67336-19-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +126 to +140 deg (C=1, 1N HCl) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...