4-(3-Nitrophenyl)-1,3-thiazol-2-amine ≥98.0% (by GC, titration analysis)
Proveedor: TCI
|
Atención
|
A3086-1G
A3086-200MG
TCIAA3086-1GEA
107
EUR
TCIAA3086-1G
TCIAA3086-200MG
4-(3-Nitrophenyl)-1,3-thiazol-2-amine ≥98.0% (by GC, titration analysis)
4-(3-Nitrophenyl)-1,3-thiazol-2-amine
Formula:
C₉H₇N₃O₂S |
Número MDL:
MFCD00022455 Núm. CAS: 57493-24-0 |
Resultados test especificaciones
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 187.0 to 191.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...