5-Azoniaspiro[4.4]nonane Hexafluorophosphate ≥96.0% (by total nitrogen basis)

Proveedor: TCI
Atención

A3102-1G A3102-5G
TCIAA3102-1GEA 53 EUR
TCIAA3102-1G TCIAA3102-5G
5-Azoniaspiro[4.4]nonane Hexafluorophosphate ≥96.0% (by total nitrogen basis)
5-Azoniaspiro[4.4]nonane Hexafluorophosphate
Formula: C₈H₁₆F₆NP
MW: 271,19 g/mol
Núm. CAS: 129211-50-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1a13cfd9090d7f62ac0ed3b65d85df06d717b378&vid=1a13cfd9090d7f62ac0ed3b65d85df06d717b378&prid=web&clid=SAPDAM

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR