(-)-Arctigenin ≥95.0% (por GC)
Proveedor: TCI
Sinónimos:
Arctigenin, (3R,4R)-4-[(3,4-Dimethoxyphenyl)methyl]dihydro-3-[(4-hydroxy-3-methoxyphenyl)methyl]-2(3H)-furanone, (3R,4R)-4-(3,4-Dimethoxybenzyl)-3-(4-hydroxy-3-methoxybenzyl)dihydrofuran-2(3H)-one
A3117-10MG
A3117-50MG
TCIAA3117-10MGEA
60
EUR
TCIAA3117-10MG
TCIAA3117-50MG
(-)-Arctigenin ≥95.0% (por GC)
(-)-Arctigenin
Formula:
C₂₁H₂₄O₆ MW: 372,4 g/mol |
Número MDL:
MFCD00870597 Núm. CAS: 7770-78-7 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Specific rotation [a]20/D | -40.0 to -44.0 deg (C=1, methanol) |
Purity (GC) | min. 95.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...