2-Amino-4'-bromobenzophenone ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: (2-Aminophenyl)(4-bromophenyl)methanone, 2-Aminophenyl (4-bromophenyl)methanone, 2-Amino-4′-bromobenzophenone

A3162-1G A3162-5G
TCIAA3162-1GEA 33 EUR
TCIAA3162-1G TCIAA3162-5G
2-Amino-4'-bromobenzophenone ≥98.0% (by titrimetric analysis)
2-Amino-4'-bromobenzophenone
Formula: C₁₃H₁₀BrNO
MW: 276,1300 g/mol
Número MDL: MFCD02181034
Núm. CAS: 1140-17-6

Order Now

Resultados test especificaciones

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 109.0 to 113.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR