5-Amino-3,4-dimetilisoxazol ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

A3179-5G A3179-25G
TCIAA3179-5GEA 117 EUR
TCIAA3179-5G TCIAA3179-25G
5-Amino-3,4-dimetilisoxazol ≥98.0% (by GC, titration analysis)
5-Amino-3,4-dimetilisoxazol
Formula: C₅H₈N₂O
Número MDL: MFCD00046081
Núm. CAS: 19947-75-2
EINECS: 243-437-4

Order Now

Resultados test especificaciones

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR