Ac-β-Ala-OH ≥98.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: N-Ac-β-Ala-OH, 3-Acetamidopropanoic acid, N-Acetyl-beta-alanine, N-Acetyl-b-alanine

A3205-5G A3205-25G
TCIAA3205-5GEA 42 EUR
TCIAA3205-5G TCIAA3205-25G
Ac-β-Ala-OH ≥98.0% (by titrimetric analysis)
Ac-β-Ala-OH
Formula: C₅H₉NO₃
Número MDL: MFCD00037296
Núm. CAS: 3025-95-4
EINECS: 221-185-6

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 79.0 to 83.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR