21-(Acetyloxy)-17-hydroxypregn-4-ene-3,20-dione ≥97.0% (by HPLC)
Proveedor: TCI
Sinónimos:
17-Hydroxy-3,20-dioxopregn-4-en-21-yl Acetate
A3212-1G
A3212-5G
TCIAA3212-1GEA
52
EUR
TCIAA3212-1G
TCIAA3212-5G
21-(Acetyloxy)-17-hydroxypregn-4-ene-3,20-dione ≥97.0% (by HPLC)
21-(Acetyloxy)-17-hydroxypregn-4-ene-3,20-dione
Formula:
C₂₃H₃₂O₅ MW: 388,50 g/mol |
Número MDL:
MFCD00051137 Núm. CAS: 640-87-9 EINECS: 211-369-4 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Melting point | 233.0 to 237.0 deg-C |
Specific rotation [a]20/D | +123 to +136 deg (C=0.2, Acetone) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...