Abafungin ≥95.0% (by HPLC)
Proveedor: TCI
Sinónimos:
N-[4-[2-(2,4-Dimethylphenoxy)phenyl]-2-thiazolyl]-1,4,5,6-tetrahydro-2-pyrimidinamine, 4-(2-(2,4-Dimethylphenoxy)phenyl)-N-(1,4,5,6-tetrahydropyrimidin-2-yl)thiazol-2-amine, 4-[2-(2,4-Dimethylphenoxy)phenyl]-N-(1,4,5,6-tetrahydropyrimidin-2-yl)thiazol-2-amine
A3213-250MG
A3213-50MG
TCIAA3213-250MGEA
339
EUR
TCIAA3213-250MG
TCIAA3213-50MG
Abafungin ≥95.0% (by HPLC)
Abafungin
Formula:
C₂₁H₂₂N₄OS MW: 378,49 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00942263 Núm. CAS: 129639-79-8 |
Resultados test especificaciones
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Melting point | 197.0 to 201.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...