Azetidin-3-ylmethanol hydrochloride ≥96.0% (by total nitrogen and titration analysis)

Proveedor: TCI

Sinónimos: 3-Azetidinemethanol hydrochloride

A3231-1G A3231-5G
TCIAA3231-1GEA 93 EUR
TCIAA3231-1G TCIAA3231-5G
Azetidin-3-ylmethanol hydrochloride ≥96.0% (by total nitrogen and titration analysis)
Azetidin-3-ylmethanol hydrochloride
Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
Número MDL: MFCD09743446
Núm. CAS: 928038-44-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 96.0 %
Purity (with Total Nitrogen) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR