alpha-Arbutin ≥95.0% (by HPLC)

Proveedor: TCI

Sinónimos: 4-Hydroxyphenyl alpha-D-Glucopyranoside, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-hydroxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol

A3284-5G A3284-25G
TCIAA3284-5GEA 22 EUR
TCIAA3284-5G TCIAA3284-25G
alpha-Arbutin ≥95.0% (by HPLC)
alpha-Arbutin
Formula: C₁₂H₁₆O₇
MW: 272,25 g/mol
Número MDL: MFCD09838262
Núm. CAS: 84380-01-8
EINECS: 440-470-8

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Melting point 204.0 to 208.0 deg-C
Specific rotation [a]20/D +174 to +186 deg (C=1, water)
Water max. 3.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR