2'-Anilino-6'-(dibutylamino)-3'-methylfluoran ≥98.0% (by HPLC)

Proveedor: TCI

Sinónimos: 6'-(Dibutylamino)-3'-methyl-2'-(phenylamino)-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, 3'-Dibutylamino-6'-methyl-7'-phenylaminofluoran, ODB 2

A3289-25G A3289-5G
TCIAA3289-25GEA 92 EUR
TCIAA3289-25G TCIAA3289-5G
2'-Anilino-6'-(dibutylamino)-3'-methylfluoran ≥98.0% (by HPLC)
2'-Anilino-6'-(dibutylamino)-3'-methylfluoran
Formula: C₃₅H₃₆N₂O₃
MW: 532,68 g/mol
Número MDL: MFCD00307579
Núm. CAS: 89331-94-2
EINECS: 403-830-5

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %
Melting point 183.0 to 187.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR