(S)-3-Aminobutyric acid ≥98.0% (by titrimetric analysis)

Proveedor: TCI

A3309-1G A3309-250MG
TCIAA3309-1GEA 143 EUR
TCIAA3309-1G TCIAA3309-250MG
(S)-3-Aminobutyric acid ≥98.0% (by titrimetric analysis)
(S)-3-Aminobutyric acid
Formula: C₄H₉NO₂
Número MDL: MFCD00270347
Núm. CAS: 3775-72-2

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +34.0 to +38.0 deg (C=1, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR