4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Proveedor: TCI
Sinónimos:
(1,1-Dioxidotetrahydro-2H-thiopyran-4-yl)amine Hydrochloride, 4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride hydrochloride, (1,1-Dioxidotetrahydro-2H-thiopyran-4-yl)amine hydrochloride
A3327-1G
A3327-200MG
TCIAA3327-1GEA
90
EUR
TCIAA3327-1G
TCIAA3327-200MG
4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride ≥98.0% (by total nitrogen and titration analysis)
4-Aminotetrahydro-2H-thiopyran 1,1-dioxide hydrochloride
Formula:
C₅H₁₂ClNO₂S MW: 185,6700 g/mol Pto. de fusión: 289°C |
Número MDL:
MFCD11114559 Núm. CAS: 116529-31-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...