2-Amino-3-bromo-5-fluoropiridina ≥98.0% (by GC, titration analysis)

Proveedor: TCI
Atención

A3346-1G A3346-5G
TCIAA3346-1GEA 50 EUR
TCIAA3346-1G TCIAA3346-5G
2-Amino-3-bromo-5-fluoropiridina ≥98.0% (by GC, titration analysis)
2-Amino-3-bromo-5-fluoropiridina
Formula: C₅H₄BrFN₂
Número MDL: MFCD03092924
Núm. CAS: 869557-43-7

Order Now

Resultados test especificaciones

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 64.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR