Alloc-Val-Cit-PAB-OH ≥96.0% (by HPLC, total nitrogen)
Proveedor: TCI
Sinónimos:
N-(Allyloxycarbonyl)-L-valyl-N5-(aminocarbonyl)-N-[4-(hydroxymethyl)phenyl]-L-ornithinamide, Allyl [(S)-1-[[(S)-1-[[4-(Hydroxymethyl)phenyl]amino]-1-oxo-5-ureidopentan-2-yl]amino]-3-methyl-1-oxobutan-2-yl]carbamate
A3348-1G
A3348-250MG
TCIAA3348-1GEA
859
EUR
TCIAA3348-1G
TCIAA3348-250MG
Alloc-Val-Cit-PAB-OH ≥96.0% (by HPLC, total nitrogen)
Alloc-Val-Cit-PAB-OH
Formula:
C₂₂H₃₃N₅O₆ MW: 463,54 g/mol Temperatura de almacenaje: Congelador |
Núm. CAS:
1608127-09-8 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (with Total Nitrogen) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...