9-Acetylcarbazole ≥98.0% (por GC)
Proveedor: TCI
A3373-1G
A3373-5G
TCIAA3373-1GEA
25
EUR
TCIAA3373-1G
TCIAA3373-5G
9-Acetylcarbazole ≥98.0% (por GC)
9-Acetylcarbazole
Formula:
C₁₄H₁₁NO |
Número MDL:
MFCD00087027 Núm. CAS: 574-39-0 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 75.0 to 79.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...