Bis(2,4-dinitrophenyl) oxalate ≥98.0% (by titrimetric analysis) chemiluminescence reagent

Proveedor: TCI
Peligro

Sinónimos: Oxalic Acid Bis(2,4-dinitrophenyl) Ester, DNPO

A5303-1G A5303-5G
TCIAA5303-1GEA 43 EUR
TCIAA5303-1G TCIAA5303-5G
Bis(2,4-dinitrophenyl) oxalate ≥98.0% (by titrimetric analysis) chemiluminescence reagent
Bis(2,4-dinitrophenyl) oxalate

Chemiluminescence reagent for the determination of fluorescent compounds by HPLC and FIA

Formula: C₁₄H₆N₄O₁₂
MW: 422,22 g/mol
Número MDL: MFCD00010712
Núm. CAS: 16536-30-4
UN: 1325
ADR: 4.1,II

Order Now

Resultados test especificaciones

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR