ABD-F [=4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole] ≥98.0% (by total nitrogen and titration analysis) HPLC labeling reagent for determination of thiols
Proveedor: TCI
|
Atención
|
Sinónimos:
ABD-F, 4-Fluoro-7-sulfobenzofurazan, ammonium salt, 7-Fluoro-2,1,3-Benzoxadiazole-4-sulphonamide, 4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole, 4-(Aminosulfonyl)-7-fluorobenzofurazane, 7-Fluoro-2,1,3-benzoxadiazole-4-sulfonamide, 7-Fluorobenzofurazan-4-sulfonamide
A5597-100MG
A5597-500MG
TCIAA5597-100MGEA
214
EUR
TCIAA5597-100MG
TCIAA5597-500MG
ABD-F [=4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole] ≥98.0% (by total nitrogen and titration analysis) HPLC labeling reagent for determination of thiols
ABD-F [=4-(Aminosulphonyl)-7-fluoro-2,1,3-benzoxadiazole]
Formula:
C₆H₄FN₃O₃S MW: 217,17 g/mol |
Número MDL:
MFCD00036687 Núm. CAS: 91366-65-3 |
Resultados test especificaciones
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 144.0 to 148.0 deg-C |
Effect test of HPLC derivatization | Effective as labeling agent for N-Acetyl-L-cysteine |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...