Phenyl phosphate ≥99.0% (by titrimetric analysis)

Proveedor: TCI
Atención

Sinónimos: Phosphoric Acid Phenyl Ester, Dihydrogen Phenyl Phosphate

B0026.0025 B0026-25G B0026-250G
TCIAB0026.0025EA 0 EUR
TCIAB0026.0025 TCIAB0026-25G TCIAB0026-250G
Phenyl phosphate ≥99.0% (by titrimetric analysis)
Phenyl phosphate
Formula: C₆H₇O₄P
MW: 174,09 g/mol
Número MDL: MFCD00058970
Núm. CAS: 701-64-4
EINECS: 211-857-7
UN: 3077
ADR: 9,III

Order Now

Resultados test especificaciones

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 99.0 %
Melting point 98.0 to 103.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR