N-Benzoyl-L-glutamic acid ≥98.0% (by HPLC, titration analysis)
Proveedor: TCI
B0202-1G
B0202-25G
TCIAB0202-1GEA
16
EUR
TCIAB0202-1G
TCIAB0202-25G
N-Benzoyl-L-glutamic acid ≥98.0% (by HPLC, titration analysis)
N-Benzoyl-L-glutamic acid
Formula:
C₁₂H₁₃NO₅ MW: 251,24 g/mol |
Número MDL:
MFCD00014028 Núm. CAS: 6094-36-6 |
Resultados test especificaciones
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 136.0 to 144.0 deg-C |
Specific rotation [a]20/D | +16.0 to +21.0 deg (C=10, 1mol/L NaOH sol.) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...